Product Details

SNP ID
rs12543263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:132123663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATTTATTCTGTGACTCAATCAC[C/T]GCTTCAATCACTGAGGGACGTGGTG
Phenotype
MIM: 602232
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNQ3 PubMed Links

Gene Details

Gene
KCNQ3
Gene Name
potassium voltage-gated channel subfamily Q member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204824.1 10977 UTR 3 NP_001191753.1
NM_004519.3 10977 UTR 3 NP_004510.1
XM_005250914.3 10977 UTR 3 XP_005250971.1
XM_006716555.3 10977 UTR 3 XP_006716618.1
XM_011517026.2 10977 UTR 3 XP_011515328.1
XM_017013400.1 10977 UTR 3 XP_016868889.1

View Full Product Details