Product Details

SNP ID
rs2436130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:132123330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTGGAGGGGAGGAGTGCCTGCA[G/T]GGCCATGGGATTGTTCTTCATTCCC
Phenotype
MIM: 602232
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNQ3 PubMed Links

Gene Details

Gene
KCNQ3
Gene Name
potassium voltage-gated channel subfamily Q member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204824.1 11310 UTR 3 NP_001191753.1
NM_004519.3 11310 UTR 3 NP_004510.1
XM_005250914.3 11310 UTR 3 XP_005250971.1
XM_006716555.3 11310 UTR 3 XP_006716618.1
XM_011517026.2 11310 UTR 3 XP_011515328.1
XM_017013400.1 11310 UTR 3 XP_016868889.1

View Full Product Details