Product Details

SNP ID
rs1284371
Assay Type
Validated
NCBI dbSNP Submissions
35
Location
Chr.1:32887170 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGCACCAACTTACAGAAACATGCA[A/T]GTGACACACCGCCGAGGACACACAC
Phenotype
MIM: 142622
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HPCA PubMed Links

Gene Details

Gene
HPCA
Gene Name
hippocalcin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002143.2 98 Intron NP_002134.2
XM_005270792.2 98 Intron XP_005270849.1
XM_017001118.1 98 UTR 5 XP_016856607.1
Gene
TMEM54
Gene Name
transmembrane protein 54
There are no transcripts associated with this gene.

View Full Product Details