Product Details

SNP ID
rs2147064
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:86278976 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCACCAACAGGATCTACTCTCA[C/T]GTACTTAACCGTTCTTTCACTGTTA
Phenotype
MIM: 611006 MIM: 613467
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ISCA1 PubMed Links

Gene Details

Gene
ISCA1
Gene Name
iron-sulfur cluster assembly 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030940.3 Intron NP_112202.2
Gene
ZCCHC6
Gene Name
zinc finger CCHC-type containing 6
There are no transcripts associated with this gene.

View Full Product Details