Product Details

SNP ID
rs1055409
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:87085592 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGAACTAGACTGCTGATTGTACTGT[A/G]GGATAAGGAGGAAGGCAGTAAAGCT
Phenotype
MIM: 118850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CGA PubMed Links

Gene Details

Gene
CGA
Gene Name
glycoprotein hormones, alpha polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000735.3 750 UTR 3 NP_000726.1
NM_001252383.1 750 UTR 3 NP_001239312.1

View Full Product Details