Product Details

SNP ID
rs34655625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:11926861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAATTGGAGGGAGGAAGGGAAAA[C/T]AGAAAGTGAAGGAACTCTGCAAAGT
Phenotype
MIM: 605529
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UPF2 PubMed Links

Gene Details

Gene
UPF2
Gene Name
UPF2 regulator of nonsense transcripts homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015542.3 Intron NP_056357.1
NM_080599.2 Intron NP_542166.1
XM_011519449.2 Intron XP_011517751.1
XM_017016034.1 Intron XP_016871523.1

View Full Product Details