Product Details

SNP ID
rs12496524
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:126526007 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CCCAGCCTCTGCGCAGGCTGGGCTC[C/T]CCACCTGGCCTGTCCTGCTCTTGGC
Phenotype
MIM: 610124 MIM: 613012
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C3orf22 PubMed Links

Gene Details

Gene
C3orf22
Gene Name
chromosome 3 open reading frame 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152533.2 4031 Intron NP_689746.1
XM_011512455.2 4031 Intron XP_011510757.1
XM_011512457.2 4031 Intron XP_011510759.1
XM_017005751.1 4031 Intron XP_016861240.1
XM_017005752.1 4031 Intron XP_016861241.1
XM_017005753.1 4031 UTR 3 XP_016861242.1
Gene
CHST13
Gene Name
carbohydrate sulfotransferase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152889.2 4031 Intron NP_690849.1
Gene
UROC1
Gene Name
urocanate hydratase 1
There are no transcripts associated with this gene.

View Full Product Details