Product Details
- SNP ID
-
rs4135050
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:103968698 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGAGAGGCCAGATTAAAGCAGTGTT[A/T]GAGGATTTGTAGCAAATGTAAGGGA
- Phenotype
-
MIM: 601423
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C12orf73
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4135051] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C12orf73
- Gene Name
- chromosome 12 open reading frame 73
There are no transcripts associated with this gene.
- Gene
- TDG
- Gene Name
- thymine DNA glycosylase
View Full Product Details