Product Details

SNP ID
rs1384803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:78669393 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATATGTAAAACATTTAGCTAAGT[A/G]CCTAGTGCATAGGAGGCCCTCTCAG
Phenotype
MIM: 146660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL7 PubMed Links

Gene Details

Gene
IL7
Gene Name
interleukin 7
There are no transcripts associated with this gene.

Gene
ZC2HC1A
Gene Name
zinc finger C2HC-type containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016010.2 Intron NP_057094.2
XM_005251256.1 Intron XP_005251313.1
XM_006716454.2 Intron XP_006716517.1
XM_011517539.2 Intron XP_011515841.1

View Full Product Details