Product Details

SNP ID
rs2282439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125840566 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTACGAGTCTGTGTCCCCCGTCACC[A/G]TGCCCCAAGCAAGCTTGCTCTGAAG
Phenotype
MIM: 611883 MIM: 607960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCCIP PubMed Links

Gene Details

Gene
BCCIP
Gene Name
BRCA2 and CDKN1A interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016567.3 Intron NP_057651.1
NM_078468.2 Intron NP_510868.1
NM_078469.2 Intron NP_510869.1
Gene
DHX32
Gene Name
DEAH-box helicase 32 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018180.2 Intron NP_060650.2
XM_017016404.1 Intron XP_016871893.1
XM_017016405.1 Intron XP_016871894.1

View Full Product Details