Product Details

SNP ID
rs2307117
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:58491549 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGACACACCAATCTTGAGGTAAGTA[A/G]GAACCTGATTTAAGGTATTTGGATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAPT PubMed Links

Gene Details

Gene
GAPT
Gene Name
GRB2-binding adaptor protein, transmembrane
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304428.1 Intron NP_001291357.1
NM_001304429.1 Intron NP_001291358.1
NM_001304431.1 Intron NP_001291360.1
NM_152687.3 Intron NP_689900.1

View Full Product Details