Product Details

SNP ID
rs17625090
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:25237936 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGCTACGCACACTACCAACATGGA[A/G]CTGTACTTAGCATTAGAAATCTATA
Phenotype
MIM: 603750
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AQP8 PubMed Links

Gene Details

Gene
AQP8
Gene Name
aquaporin 8
There are no transcripts associated with this gene.

Gene
ZKSCAN2
Gene Name
zinc finger with KRAB and SCAN domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012981.4 5191 UTR 3 NP_001012999.3
XM_017023200.1 5191 UTR 3 XP_016878689.1

View Full Product Details