Product Details

SNP ID
rs34788087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:74952159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAAAAGGCAAGCCCCTTCGCCC[C/T]TCAGCCCACCCGCCCCCAGCCCCTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf40 PubMed Links

Gene Details

Gene
C9orf40
Gene Name
chromosome 9 open reading frame 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017998.2 Intron NP_060468.2
Gene
C9orf41-AS1
Gene Name
C9orf41 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details