Product Details

SNP ID
rs2441066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:26756799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACAACAAGTACATATTATTCTC[A/T]TTATAATTCTGACTATTGGAGATAT
Phenotype
MIM: 600752
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR12 PubMed Links

Gene Details

Gene
GPR12
Gene Name
G protein-coupled receptor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005288.3 3251 UTR 3 NP_005279.1
XM_005266360.3 3251 UTR 3 XP_005266417.1

View Full Product Details