Product Details

SNP ID
rs12444102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89175007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACAAAGAGCCAGCTTTGAGAA[C/T]GCCCCCGAGAGCCGGAAGTGGGCCG
Phenotype
MIM: 114019
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH15 PubMed Links

Gene Details

Gene
CDH15
Gene Name
cadherin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004933.2 Intron NP_004924.1
Gene
LOC400558
Gene Name
uncharacterized LOC400558
There are no transcripts associated with this gene.

View Full Product Details