Product Details

SNP ID
rs10777716
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:95663611 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTAAGGTTTCAGTTTTCTTATCAGC[C/T]TTCTTTGGCCTTGGCTGTTGAACTT
Phenotype
MIM: 610401
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NTN4 PubMed Links

Gene Details

Gene
NTN4
Gene Name
netrin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021229.3 Intron NP_067052.2
Gene
PGAM1P5
Gene Name
phosphoglycerate mutase 1 pseudogene 5
There are no transcripts associated with this gene.

View Full Product Details