Product Details

SNP ID
rs15397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43230932 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATCCACACATACCCCCATCCCA[A/G]TCCCCCAGATTTTCTTCAACATGCC
Phenotype
MIM: 611800 MIM: 612053
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01126 PubMed Links

Gene Details

Gene
LINC01126
Gene Name
long intergenic non-protein coding RNA 1126
There are no transcripts associated with this gene.

Gene
THADA
Gene Name
THADA, armadillo repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083953.1 6230 UTR 3 NP_001077422.1
NM_001271643.1 6230 Intron NP_001258572.1
NM_001271644.1 6230 Intron NP_001258573.1
NM_022065.4 6230 UTR 3 NP_071348.3
XM_006712061.3 6230 UTR 3 XP_006712124.1
XM_006712062.1 6230 UTR 3 XP_006712125.1
XM_006712063.1 6230 UTR 3 XP_006712126.1
XM_006712064.2 6230 UTR 3 XP_006712127.1
XM_006712065.1 6230 UTR 3 XP_006712128.1
XM_006712066.1 6230 UTR 3 XP_006712129.1
XM_006712067.1 6230 UTR 3 XP_006712130.1
XM_006712068.3 6230 Intron XP_006712131.1
XM_006712069.3 6230 UTR 3 XP_006712132.1
XM_011533015.2 6230 Intron XP_011531317.1
XM_017004674.1 6230 UTR 3 XP_016860163.1
XM_017004675.1 6230 UTR 3 XP_016860164.1
Gene
ZFP36L2
Gene Name
ZFP36 ring finger protein like 2
There are no transcripts associated with this gene.

View Full Product Details