Product Details
- SNP ID
-
rs1549562
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:40397138 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTGATTTACGTTTCAAATACAGAC[A/T]CTCATCATTTTCAGTAGGTTATAGG
- Phenotype
-
MIM: 612170
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MUC19
PubMed Links
Gene Details
- Gene
- MUC19
- Gene Name
- mucin 19, oligomeric
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173600.2 |
|
Intron |
|
|
NP_775871.2 |
View Full Product Details