Product Details
- SNP ID
-
rs11683062
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:229948577 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTCCCTTGCAGCTACAGTTAGCC[A/G]CATGACTAAATTCTGGCCAAGGGAA
- Phenotype
-
MIM: 609105
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO36
PubMed Links
Gene Details
- Gene
- FBXO36
- Gene Name
- F-box protein 36
View Full Product Details