Product Details

SNP ID
rs4733951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38512313 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAGCCACTCTTCAGAGAGGAATA[A/C]GAAGGAATTCCTCCGCGAGGGAGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C8orf86 PubMed Links

Gene Details

Gene
C8orf86
Gene Name
chromosome 8 open reading frame 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303531.1 771 UTR 3 NP_001290460.1
NM_207412.2 771 UTR 3 NP_997295.1

View Full Product Details