Product Details
- SNP ID
-
rs6867870
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:140262311 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGCATAAGGAATCCCCATTTCTTTC[A/C]GAGTAAAAGTGAATGGTAAAGGGTT
- Phenotype
-
MIM: 604897
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PFDN1
PubMed Links
Gene Details
- Gene
- PFDN1
- Gene Name
- prefoldin subunit 1
View Full Product Details