Product Details

SNP ID
rs3729604
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:159917454 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATCTCCATCGGGCCTCTCCTTGG[A/G]TGGAAGGAGCCGGCACCCAACGATG
Phenotype
MIM: 104220
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ADRA1B PubMed Links

Gene Details

Gene
ADRA1B
Gene Name
adrenoceptor alpha 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000679.3 722 Silent Mutation GGA,GGG G183G NP_000670.1
XM_005265818.3 722 Silent Mutation GGA,GGG G183G XP_005265875.1
XM_005265819.2 722 Silent Mutation GGA,GGG G183G XP_005265876.1
XM_006714821.3 722 Silent Mutation GGA,GGG G183G XP_006714884.1
XM_011534435.1 722 Silent Mutation GGA,GGG G183G XP_011532737.1
XM_011534437.2 722 Silent Mutation GGA,GGG G183G XP_011532739.1
XM_011534438.2 722 Silent Mutation GGA,GGG G183G XP_011532740.1

View Full Product Details