Product Details

SNP ID
rs2241778
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:103400707 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAAGTAATGTAGTTTATAAGCCAT[C/T]TGTAGGGATTTACATTCCTTTTATG
Phenotype
MIM: 610815
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A32 PubMed Links

Gene Details

Gene
SLC25A32
Gene Name
solute carrier family 25 member 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030780.4 Intron NP_110407.2
XM_017013877.1 Intron XP_016869366.1

View Full Product Details