Product Details
- SNP ID
-
rs3770098
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:85578244 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGTCAAGCGGGGAGGAGCTAAATA[A/C]TATTTTTCTCTCCTTGTTCAAAAAC
- Phenotype
-
MIM: 607029
MIM: 603177
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
VAMP5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139977225] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- VAMP5
- Gene Name
- vesicle associated membrane protein 5
There are no transcripts associated with this gene.
- Gene
- VAMP8
- Gene Name
- vesicle associated membrane protein 8
View Full Product Details