Product Details

SNP ID
rs1594842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:39270120 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAGTTTTCTGCATTCTCTCCAAC[A/G]CTTGTGATTTTCTGTTTTCTTTTGT
Phenotype
MIM: 602132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTAGE5 PubMed Links

Gene Details

Gene
CTAGE5
Gene Name
CTAGE family member 5, ER export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001247988.1 Intron NP_001234917.1
NM_001247989.1 Intron NP_001234918.1
NM_001247990.1 Intron NP_001234919.1
NM_005930.3 Intron NP_005921.2
NM_203354.2 Intron NP_976229.1
NM_203355.2 Intron NP_976230.1
NM_203356.2 Intron NP_976231.1
XM_005267648.4 Intron XP_005267705.1
XM_006720148.2 Intron XP_006720211.1
XM_011536775.2 Intron XP_011535077.1
XM_011536778.2 Intron XP_011535080.1
XM_011536784.2 Intron XP_011535086.1
XM_011536785.2 Intron XP_011535087.1
XM_017021314.1 Intron XP_016876803.1
XM_017021315.1 Intron XP_016876804.1
XM_017021316.1 Intron XP_016876805.1
XM_017021317.1 Intron XP_016876806.1
XM_017021318.1 Intron XP_016876807.1
XM_017021319.1 Intron XP_016876808.1
XM_017021320.1 Intron XP_016876809.1
XM_017021321.1 Intron XP_016876810.1
XM_017021322.1 Intron XP_016876811.1
XM_017021323.1 Intron XP_016876812.1
XM_017021324.1 Intron XP_016876813.1
XM_017021325.1 Intron XP_016876814.1
XM_017021326.1 Intron XP_016876815.1
XM_017021327.1 Intron XP_016876816.1
XM_017021328.1 Intron XP_016876817.1
XM_017021329.1 Intron XP_016876818.1
XM_017021330.1 Intron XP_016876819.1
Gene
LOC100288846
Gene Name
uncharacterized LOC100288846
There are no transcripts associated with this gene.

View Full Product Details