Product Details

SNP ID
rs3111751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:230871392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCGATCCCTGGGAGATGCCAAC[A/G]GAAGGTGTCACCCCCGTCTCTTGCT
Phenotype
MIM: 609554
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITM2C PubMed Links

Gene Details

Gene
ITM2C
Gene Name
integral membrane protein 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012514.2 Intron NP_001012532.1
NM_001012516.2 Intron NP_001012534.1
NM_001287240.1 Intron NP_001274169.1
NM_001287241.1 Intron NP_001274170.1
NM_030926.5 Intron NP_112188.1

View Full Product Details