Product Details

SNP ID
rs12608688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1944798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCACTGCGGGCAGCCCTGCAGC[C/G]GGCAGGGATTGAGTGAGGGGTGGGG
Phenotype
MIM: 602214
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CSNK1G2 PubMed Links

Gene Details

Gene
CSNK1G2
Gene Name
casein kinase 1 gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319.6 Intron NP_001310.3
XM_005259498.1 Intron XP_005259555.1
XM_005259499.3 Intron XP_005259556.1
XM_005259500.1 Intron XP_005259557.1
XM_005259501.2 Intron XP_005259558.1
XM_017026297.1 Intron XP_016881786.1
XM_017026298.1 Intron XP_016881787.1
Gene
CSNK1G2-AS1
Gene Name
CSNK1G2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details