Product Details
- SNP ID
-
rs9384983
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:116637490 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTTATACATTGTTAGCCAAGTTAC[A/G]ATTCTTACAGTTAAAGCACAGCTAC
- Phenotype
-
MIM: 612647
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RSPH4A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139721376] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RSPH4A
- Gene Name
- radial spoke head 4 homolog A
There are no transcripts associated with this gene.
- Gene
- ZUFSP
- Gene Name
- zinc finger with UFM1 specific peptidase domain
View Full Product Details