Product Details
- SNP ID
-
rs9143
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:105173833 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAAGAAATGCTATTAGGTCCAATT[A/G]TGAGATAAGCTGAATTATAAATACA
- Phenotype
-
MIM: 606231
MIM: 615748
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APPL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77220426] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APPL2
- Gene Name
- adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
- Gene
- KIAA1033
- Gene Name
- KIAA1033
There are no transcripts associated with this gene.
View Full Product Details