Product Details
- SNP ID
-
rs7747099
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:36402477 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAAGAGTCCAGTGGGAGAGAGACA[C/T]AAACTACAATTCAACATGTTAGAGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PXT1
PubMed Links
Gene Details
- Gene
- PXT1
- Gene Name
- peroxisomal, testis specific 1
View Full Product Details