Product Details

SNP ID
rs11878315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:280299 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTAGGCTCAGGCCTGATAGGAACA[T/C]CCTGCAAGGGCCGGGGAATAAGATA
Phenotype
MIM: 607126
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
PLPP2 PubMed Links

Gene Details

Gene
PLPP2
Gene Name
phospholipid phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003712.3 Intron NP_003703.1
NM_177526.2 Intron NP_803545.1
NM_177543.2 Intron NP_808211.1
XM_011528396.2 Intron XP_011526698.1

View Full Product Details