Product Details

SNP ID
rs10208532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:167961916 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGGCTCTCTGGGGTTAATTTGCC[C/T]AGGATCACACAGTGGAGGCTGCAGA
Phenotype
MIM: 607648
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STK39 PubMed Links

Gene Details

Gene
STK39
Gene Name
serine/threonine kinase 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013233.2 3100 Intron NP_037365.2
XM_005246465.2 3100 Intron XP_005246522.1
XM_017003813.1 3100 UTR 3 XP_016859302.1
XM_017003814.1 3100 Intron XP_016859303.1
XM_017003815.1 3100 Intron XP_016859304.1
XM_017003816.1 3100 Intron XP_016859305.1
XM_017003817.1 3100 Intron XP_016859306.1

View Full Product Details