Product Details

SNP ID
rs6747042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169646989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTCTCAAAATCCAAAATGTTG[A/G]CATAAAATAGTAACCCAAACCTGAT
Phenotype
MIM: 606093
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC173 PubMed Links

Gene Details

Gene
CCDC173
Gene Name
coiled-coil domain containing 173
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085447.1 Intron NP_001078916.1
XM_011510589.1 Intron XP_011508891.1
XM_011510590.1 Intron XP_011508892.1
Gene
PPIG
Gene Name
peptidylprolyl isomerase G
There are no transcripts associated with this gene.

View Full Product Details