Product Details

SNP ID
rs11544800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:73196734 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTATACTCATCCGGCAGGTGGC[A/G]GCCGCCAGCCACTTCCGGTAAGTGG
Phenotype
MIM: 606973
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COG1 PubMed Links

Gene Details

Gene
COG1
Gene Name
component of oligomeric golgi complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018714.2 579 Silent Mutation GCA,GCG A181A NP_061184.1

View Full Product Details