Product Details
- SNP ID
-
rs11584005
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
24
- Location
-
Chr.1:35881973 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATAAGGGTCTGCACCAGCATAAGAA[A/G]CCATCAGAATGGCAGTGAATCATGT
- Phenotype
-
MIM: 606228
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AGO1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116188558] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGO1
- Gene Name
- argonaute 1, RISC catalytic component
View Full Product Details