Product Details

SNP ID
rs10775668
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41306102 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATCTCAAGAAACCACCCATTTCA[C/T]TCAATCATAAGAAGCAATTCCTCAT
Phenotype
MIM: 608617
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM3B PubMed Links
Additional Information
For this assay, SNP(s) [rs115287974] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FAM3B
Gene Name
family with sequence similarity 3 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058186.3 Intron NP_478066.3
NM_206964.1 Intron NP_996847.1
XM_011529648.2 Intron XP_011527950.2
XM_011529649.2 Intron XP_011527951.1

View Full Product Details