Product Details

SNP ID
rs1054220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44849795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCTGTATTTCACAACCGAGCCCT[A/C]GCGCCAGCTCAGCACCCTACGCTCA
Phenotype
MIM: 603784
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTTG1IP PubMed Links

Gene Details

Gene
PTTG1IP
Gene Name
pituitary tumor-transforming 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286822.1 2616 UTR 3 NP_001273751.1
NM_004339.3 2616 UTR 3 NP_004330.1

View Full Product Details