Product Details

SNP ID
rs873317
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:44850330 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGCAGCACTTTTCAGTGACTTCCCC[A/G]CACGCTGCTTTCCCTGCCACGAAGC
Phenotype
MIM: 603784
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTTG1IP PubMed Links

Gene Details

Gene
PTTG1IP
Gene Name
pituitary tumor-transforming 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286822.1 2081 UTR 3 NP_001273751.1
NM_004339.3 2081 UTR 3 NP_004330.1

View Full Product Details