Product Details

SNP ID
rs7797759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107924562 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTGGAATTGAATGCAAAATTGAC[A/C]TGATTTTAAAGAACATTTCAAAAAA
Phenotype
MIM: 238331 MIM: 150240
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DLD PubMed Links

Gene Details

Gene
DLD
Gene Name
dihydrolipoamide dehydrogenase
There are no transcripts associated with this gene.

Gene
LAMB1
Gene Name
laminin subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002291.2 Intron NP_002282.2
XM_017012201.1 Intron XP_016867690.1
XM_017012202.1 Intron XP_016867691.1

View Full Product Details