Product Details
- SNP ID
-
rs2240609
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:1948376 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGAGCTCCGTGCCTCAGGCCAGTTG[C/T]TCAGCCTCAGTTTCCTTGTCTGTAG
- Phenotype
-
MIM: 609843
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DCP1B
PubMed Links
Gene Details
- Gene
- DCP1B
- Gene Name
- decapping mRNA 1B
View Full Product Details