Product Details

SNP ID
rs374483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37016210 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTTTGGGCTGGATAATTCTTCAC[A/G]GTGGGAACTATCCTGGGCCTCGTAG
Phenotype
MIM: 609892
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIPPLY3 PubMed Links

Gene Details

Gene
RIPPLY3
Gene Name
ripply transcriptional repressor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317768.1 Intron NP_001304697.1
NM_001317777.1 Intron NP_001304706.1
NM_018962.2 Intron NP_061835.1

View Full Product Details