Product Details

SNP ID
rs714743
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:27865642 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCGTCTCTGCCTAGCTCTCAAGAG[A/G]TTCACAGCCTGTGGCAAGGCAAAGC
Phenotype
MIM: 606876
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PITPNB PubMed Links

Gene Details

Gene
PITPNB
Gene Name
phosphatidylinositol transfer protein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284277.1 Intron NP_001271206.1
NM_001284278.1 Intron NP_001271207.1
NM_012399.4 Intron NP_036531.1
XM_011530052.2 Intron XP_011528354.1
XM_017028707.1 Intron XP_016884196.1

View Full Product Details