Product Details

SNP ID
rs414624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75254972 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACTCCAAAATCCCCATAATTTAA[C/T]TAGGAAATTTATAGAACATCTTTTT
Phenotype
MIM: 611028
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM30A PubMed Links

Gene Details

Gene
TMEM30A
Gene Name
transmembrane protein 30A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143958.1 2386 UTR 3 NP_001137430.1
NM_018247.3 2386 UTR 3 NP_060717.1

View Full Product Details