Product Details

SNP ID
rs184371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:28999982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAGGTGAGTAACAAAATGTGTC[A/G]GGGCTGTAATTACGTATTTAAATGT
Phenotype
MIM: 609780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPVL PubMed Links

Gene Details

Gene
CPVL
Gene Name
carboxypeptidase, vitellogenic like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019029.2 Intron NP_061902.2
NM_031311.3 Intron NP_112601.3
XM_011515437.1 Intron XP_011513739.1
XM_017012366.1 Intron XP_016867855.1
Gene
LOC100506497
Gene Name
uncharacterized LOC100506497
There are no transcripts associated with this gene.

View Full Product Details