Product Details

SNP ID
rs760166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:39346235 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGCTCACAACTTCAATGCTAACA[C/T]GGGCGGAAAAATCACTGCAGGGTGT
Phenotype
MIM: 163920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMGN1 PubMed Links

Gene Details

Gene
HMGN1
Gene Name
high mobility group nucleosome binding domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004965.6 Intron NP_004956.5
XM_011529543.1 Intron XP_011527845.1
XM_011529544.1 Intron XP_011527846.1
XM_011529545.1 Intron XP_011527847.1
XM_011529547.1 Intron XP_011527849.1
XM_011529549.1 Intron XP_011527851.1
XM_017028332.1 Intron XP_016883821.1
XM_017028333.1 Intron XP_016883822.1
XM_017028334.1 Intron XP_016883823.1
XM_017028335.1 Intron XP_016883824.1
XM_017028336.1 Intron XP_016883825.1
XM_017028338.1 Intron XP_016883827.1

View Full Product Details