Product Details

SNP ID
rs533483
Assay Type
Functionally tested
NCBI dbSNP Submissions
59
Location
Chr.1:234629510 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGCTGTGTGTTTGAGTTCTGGCC[A/G]TTAGTGAGTGGAAGTGATGTGTCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00184 PubMed Links

Gene Details

Gene
LINC00184
Gene Name
long intergenic non-protein coding RNA 184
There are no transcripts associated with this gene.

View Full Product Details