Product Details
- SNP ID
-
rs741859
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:105141642 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCACGTGGGCCAATACCCAGCACG[C/T]GCCCCACTGCCCCAAGCCTCTCTGC
- Phenotype
-
MIM: 602570
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
JAG2
PubMed Links
Gene Details
- Gene
- JAG2
- Gene Name
- jagged 2
- Gene
- MIR6765
- Gene Name
- microRNA 6765
There are no transcripts associated with this gene.
View Full Product Details