Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001278370.1 | 1924 | UTR 3 | NP_001265299.1 | ||
NM_001278371.1 | 1924 | UTR 3 | NP_001265300.1 | ||
NM_001278372.1 | 1924 | UTR 3 | NP_001265301.1 | ||
NM_001278373.1 | 1924 | UTR 3 | NP_001265302.1 | ||
NM_001278374.1 | 1924 | UTR 3 | NP_001265303.1 | ||
NM_001278375.1 | 1924 | UTR 3 | NP_001265304.1 | ||
NM_001278376.2 | 1924 | UTR 3 | NP_001265305.2 | ||
NM_001278381.1 | 1924 | UTR 3 | NP_001265310.1 | ||
NM_005374.4 | 1924 | UTR 3 | NP_005365.4 | ||
XM_011524827.2 | 1924 | UTR 3 | XP_011523129.2 |