Product Details
- SNP ID
-
rs35405
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:33945653 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAATACTAGCTATCACATTTTGAGT[C/T]TTTGTCGGGTGCTAGGGGGCACTAT
- Phenotype
-
MIM: 609445
MIM: 606202
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RXFP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs59234689] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RXFP3
- Gene Name
- relaxin/insulin like family peptide receptor 3
There are no transcripts associated with this gene.
- Gene
- SLC45A2
- Gene Name
- solute carrier family 45 member 2
View Full Product Details