Product Details

SNP ID
rs37362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58165013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTATTTGAAACACACGTTATCAG[A/T]GGAAAGAACCATACACATGCAGAAG
Phenotype
MIM: 115442
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CSNK2A2 PubMed Links

Gene Details

Gene
CSNK2A2
Gene Name
casein kinase 2 alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001896.2 Intron NP_001887.1
XM_005255800.3 Intron XP_005255857.1
XM_005255801.3 Intron XP_005255858.1
XM_017022945.1 Intron XP_016878434.1

View Full Product Details